Learn More
Medchemexpress LLC Bromo-PEG3-bromide | 31255-26-2 | 98.8% | C8H16Br2O3 | 250 MG
SDP
Catalog No. 5000352671
Encompass_Preferred
Bromo-PEG3-bromide is a PEG-based PROTAC linker. It is utilized in the synthesis of PROTACs, which are compounds designed to exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is for research use only.
  • PEG-based PROTAC linker
  • Used in PROTAC synthesis
  • Contains two distinct ligands connected by a linker
  • One ligand targets an E3 ubiquitin ligase
  • The other ligand targets a protein of interest
  • Leverages the ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-52671 Supplier Medchemexpress LLC Supplier No. 250MGHY141373
Bromo-PEG3-bromide is a PEG-based PROTAC linker. It is utilized in the synthesis of PROTACs, which are compounds designed to exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is for research use only.
  • PEG-based PROTAC linker
  • Used in PROTAC synthesis
  • Contains two distinct ligands connected by a linker
  • One ligand targets an E3 ubiquitin ligase
  • The other ligand targets a protein of interest
  • Leverages the ubiquitin-proteasome system
  • Selectively degrades target proteins