Learn More
Medchemexpress LLC Bromo-PEG3-bromide | 31255-26-2 | MFCD18205853 | 98.8% | C8H16Br2O3 | 5 G
SDP
Catalog No. 5000355723
Encompass_Preferred
Bromo-PEG3-bromide is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins
Catalog No. 50-003-55723 Supplier Medchemexpress LLC Supplier No. HY1413735G
Bromo-PEG3-bromide is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins