Learn More
Medchemexpress LLC Bromo-PEG4-bromide | 57602-02-5 | 98.0% | 364.07 | C10H20Br2O4 | 1 G
SDP
Catalog No. 5000208937
Encompass_Preferred
Bromo-PEG4-bromide is a bromo-terminated polyethylene glycol (PEG4) linker used in the synthesis of PROTACs and other bioconjugation applications. It is supplied as a research-grade reagent with documented purity and handling information.
  • Bromo-terminated PEG4 spacer for linker installation.
  • High purity suitable for synthetic chemistry (98.0%).
  • Available in multiple pack sizes; recommended storage at -20°C for long-term stability.
  • Provided with datasheet, SDS, COA, and spectral data for quality verification.
Catalog No. 50-002-08937 Supplier Medchemexpress LLC Supplier No. HYW0405271G
Bromo-PEG4-bromide is a bromo-terminated polyethylene glycol (PEG4) linker used in the synthesis of PROTACs and other bioconjugation applications. It is supplied as a research-grade reagent with documented purity and handling information.
  • Bromo-terminated PEG4 spacer for linker installation.
  • High purity suitable for synthetic chemistry (98.0%).
  • Available in multiple pack sizes; recommended storage at -20°C for long-term stability.
  • Provided with datasheet, SDS, COA, and spectral data for quality verification.