Learn More
Medchemexpress LLC Bromo-PEG6-azide | 2062663-64-1 | 95.0% | C14H28BrN3O6 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY140823100MG
Encompass_Preferred
Bromo-PEG6-azide is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, possessing an azide group that facilitates copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with alkyne-containing molecules. Additionally, it can engage in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Click chemistry reagent
  • Facilitates copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc)
  • Enables strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
Catalog No. 50-003-53163