Learn More
Medchemexpress LLC Bromo-PEG6-Boc | 1393330-41-0 | 97.0% | C19H37BrO8 | 250 MG
SDP
Catalog No. 5000352834
Encompass_Preferred
Bromo-PEG6-Boc is a PEG-based PROTAC linker used in the synthesis of PROTACs.
  • PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein.
  • PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
Catalog No. 50-003-52834 Supplier Medchemexpress LLC Supplier No. 250MGHY141367
Bromo-PEG6-Boc is a PEG-based PROTAC linker used in the synthesis of PROTACs.
  • PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein.
  • PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.