Learn More
Medchemexpress LLC Bromo-PEG8-Boc | 1623792-00-6 | 99.86% | C23H45BrO10 | 500 MG
SDP
Catalog No. 5000355318
Encompass_Preferred
Bromo-PEG8-Boc is a PEG-based PROTAC linker essential for the synthesis of Proteolysis-Targeting Chimeras (PROTACs). PROTACs are designed to contain two distinct ligands, one for an E3 ubiquitin ligase and another for a target protein, connected by a linker. This mechanism allows PROTACs to exploit the intracellular ubiquitin-proteasome system, enabling the selective degradation of specific target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Enables selective degradation of target proteins
  • Applicable in cancer research
  • Supports targeted therapy
Catalog No. 50-003-55318 Supplier Medchemexpress LLC Supplier No. HY141368500MG
Bromo-PEG8-Boc is a PEG-based PROTAC linker essential for the synthesis of Proteolysis-Targeting Chimeras (PROTACs). PROTACs are designed to contain two distinct ligands, one for an E3 ubiquitin ligase and another for a target protein, connected by a linker. This mechanism allows PROTACs to exploit the intracellular ubiquitin-proteasome system, enabling the selective degradation of specific target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Enables selective degradation of target proteins
  • Applicable in cancer research
  • Supports targeted therapy