Learn More
Medchemexpress LLC Bromo-PEG8-Boc | 1623792-00-6 | 99.9% | C23H45BrO10 | 100 MG
SDP
Catalog No. 5000354025
Encompass_Preferred
Bromo-PEG8-Boc is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. PROTACs function by leveraging the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Utilized in the synthesis of PROTACs.
  • Leverages the intracellular ubiquitin-proteasome system.
  • Facilitates selective degradation of target proteins.
Catalog No. 50-003-54025 Supplier Medchemexpress LLC Supplier No. HY141368100MG
Bromo-PEG8-Boc is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. PROTACs function by leveraging the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Utilized in the synthesis of PROTACs.
  • Leverages the intracellular ubiquitin-proteasome system.
  • Facilitates selective degradation of target proteins.