Learn More
Medchemexpress LLC Bromo-PEG8-Boc | 1623792-00-6 | 99.9% | C23H45BrO10 | 250 MG
SDP
Catalog No. 5000352559
Encompass_Preferred
Bromo-PEG8-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Used in the synthesis of PROTACs
  • Contains two different ligands connected by a linker
  • One ligand targets an E3 ubiquitin ligase
  • Other ligand targets the protein of interest
  • Facilitates selective degradation of target proteins
Catalog No. 50-003-52559 Supplier Medchemexpress LLC Supplier No. 250MGHY141368
Bromo-PEG8-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Used in the synthesis of PROTACs
  • Contains two different ligands connected by a linker
  • One ligand targets an E3 ubiquitin ligase
  • Other ligand targets the protein of interest
  • Facilitates selective degradation of target proteins