Learn More
Medchemexpress LLC Bromo-PEG8-bromide | 2376383-09-2 | 98.0% | 540.28 g/mol | C18H36Br2O8 | 1 G
SDP
Catalog No. 5000283189
Encompass_Preferred
Bromo-PEG8-bromide is a bifunctional polyethylene glycol (PEG) linker bearing terminal bromide groups on both ends. It is a hydrophilic spacer useful for nucleophilic substitution reactions and bioconjugation in drug-delivery research and linker synthesis for antibody-drug conjugates and PROTACs. Molecular formula C18H36Br2O8, molecular weight 540.28 g/mol, CAS 2376383-09-2.
  • Bifunctional PEG linker with terminal bromide groups for facile nucleophilic substitution.
  • Enhances solubility and flexibility of conjugates in aqueous media.
  • Suitable for bioconjugation and linker synthesis in drug-delivery research.
  • Useful for constructing linkers in ADC and PROTAC workflows.
  • Typically supplied as a liquid for straightforward handling.
Catalog No. 50-002-83189 Supplier Medchemexpress LLC Supplier No. HYW1908711G
Bromo-PEG8-bromide is a bifunctional polyethylene glycol (PEG) linker bearing terminal bromide groups on both ends. It is a hydrophilic spacer useful for nucleophilic substitution reactions and bioconjugation in drug-delivery research and linker synthesis for antibody-drug conjugates and PROTACs. Molecular formula C18H36Br2O8, molecular weight 540.28 g/mol, CAS 2376383-09-2.
  • Bifunctional PEG linker with terminal bromide groups for facile nucleophilic substitution.
  • Enhances solubility and flexibility of conjugates in aqueous media.
  • Suitable for bioconjugation and linker synthesis in drug-delivery research.
  • Useful for constructing linkers in ADC and PROTAC workflows.
  • Typically supplied as a liquid for straightforward handling.