Learn More
Medchemexpress LLC Bromo-PEG8-bromide | 2376383-09-2 | MFCD32861289 | 98.0% | 540.28 g/mol | C18H36Br2O8 | 500 MG
SDP
Catalog No. 5000283137
Encompass_Preferred
Bromo-PEG8-bromide is a polyethylene glycol (PEG) derivative bearing terminal bromide groups. It is used as a bromo-functionalized linker for PEGylation, bioconjugation, and drug-delivery research; the bromide termini act as good leaving groups for nucleophilic substitution, facilitating attachment to thiols, amines, and synthesis of PEG-linked probes and linkers. Supplied for research use.
Catalog No. 50-002-83137 Supplier Medchemexpress LLC Supplier No. HYW190871500MG
Bromo-PEG8-bromide is a polyethylene glycol (PEG) derivative bearing terminal bromide groups. It is used as a bromo-functionalized linker for PEGylation, bioconjugation, and drug-delivery research; the bromide termini act as good leaving groups for nucleophilic substitution, facilitating attachment to thiols, amines, and synthesis of PEG-linked probes and linkers. Supplied for research use.