Learn More
Medchemexpress LLC Bromoacetamido-C2-PEG2-NH-Boc | 182244-33-3 | C13H25BrN2O5 | 250 MG
SDP
Catalog No. 5000352878
Encompass_Preferred
Bromoacetamido-C2-PEG2-NH-Boc is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. The appearance of the product is a solid with a white to off-white color.
  • Utilized in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Solid appearance
  • White to off-white color
Catalog No. 50-003-52878 Supplier Medchemexpress LLC Supplier No. 250MGHY141199
Bromoacetamido-C2-PEG2-NH-Boc is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. The appearance of the product is a solid with a white to off-white color.
  • Utilized in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Solid appearance
  • White to off-white color