Learn More
Medchemexpress LLC Bromoacetamido-C2-PEG2-NH-Boc | 182244-33-3 | C13H25BrN2O5 | 5 G
SDP
Catalog No. 5000356262
Encompass_Preferred
Bromoacetamido-C2-PEG2-NH-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-56262 Supplier Medchemexpress LLC Supplier No. HY1411995G
Bromoacetamido-C2-PEG2-NH-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins