Learn More
Broadpharm Bromoacetamido-PEG11-azide, 1g
Catalog No. NC3963570
Encompass
Bromoacetamido-PEG11-azide is a bifunctional linker comprised of bromine (Br), which can be used as a leaving group for nucleophilic substitution reactions, and azide group, which can undergo copper-catalyzed Click Chemistry reactions with alkynes, DBCO and BCN. The water solubility of the molecule is increased by the PEG chain. Please contact us for GMP-grade inquiries.
Catalog No. NC3963570 Supplier Broadpharm Supplier No. BP416841G
Bromoacetamido-PEG11-azide is a bifunctional linker comprised of bromine (Br), which can be used as a leaving group for nucleophilic substitution reactions, and azide group, which can undergo copper-catalyzed Click Chemistry reactions with alkynes, DBCO and BCN. The water solubility of the molecule is increased by the PEG chain. Please contact us for GMP-grade inquiries.