Learn More
Medchemexpress LLC Bromoacetamido-PEG2-C2-NHS ester | 1353011-78-5 | 95.0% | C13H19BrN2O7 | 25 MG
SDP
Catalog No. 5000357268
Encompass_Preferred
Bromoacetamido-PEG2-C2-NHS ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • PROTACs contain two different ligands connected by a linker
  • Selectively degrades target proteins
Catalog No. 50-003-57268 Supplier Medchemexpress LLC Supplier No. 25MGHY141386
Bromoacetamido-PEG2-C2-NHS ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • PROTACs contain two different ligands connected by a linker
  • Selectively degrades target proteins