Learn More
Medchemexpress LLC Bromoacetamido-PEG2-C2-NHS ester | 1353011-78-5 | 95.0% | C13H19BrN2O7 | 50 MG
SDP
Catalog No. 5000356831
Encompass_Preferred
Bromoacetamido-PEG2-C2-NHS ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This compound has a molecular weight of 395.20 and appears as a white to off-white solid-liquid mixture.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Solid-liquid mixture, white to off-white in color
Catalog No. 50-003-56831 Supplier Medchemexpress LLC Supplier No. HY14138650MG
Bromoacetamido-PEG2-C2-NHS ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This compound has a molecular weight of 395.20 and appears as a white to off-white solid-liquid mixture.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Solid-liquid mixture, white to off-white in color