Learn More
Medchemexpress LLC Bromoacetamido-PEG3-C2-Boc | 1807537-33-2 | C15H28BrNO6 | 500 MG
SDP
Catalog No. 5000357012
Encompass_Preferred
Bromoacetamido-PEG3-C2-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs are molecules that contain two different ligands connected by a linker; one ligand is for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Facilitates selective degradation of target proteins
Catalog No. 50-003-57012 Supplier Medchemexpress LLC Supplier No. HY141388500MG
Bromoacetamido-PEG3-C2-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs are molecules that contain two different ligands connected by a linker; one ligand is for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Facilitates selective degradation of target proteins