Learn More
Medchemexpress LLC Bromoacetamido-PEG3-NH-Boc | 1421933-39-2 | ≥98.0% | C15H29BrN2O6 | 1 G
SDP
Catalog No. 5000353185
Encompass_Preferred
Bromoacetamido-PEG3-NH-Boc is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs feature two distinct ligands connected by a linker, one binding to an E3 ubiquitin ligase and the other to the target protein, enabling the selective degradation of target proteins through the intracellular ubiquitin-proteasome system.
  • PEG-based linker
  • Utilized in the synthesis of PROTACs
  • Connects ligands for E3 ubiquitin ligase and target proteins
  • Enables selective degradation of target proteins
  • Molecular weight: 413.30
Catalog No. 50-003-53185 Supplier Medchemexpress LLC Supplier No. HY1412001G
Bromoacetamido-PEG3-NH-Boc is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs feature two distinct ligands connected by a linker, one binding to an E3 ubiquitin ligase and the other to the target protein, enabling the selective degradation of target proteins through the intracellular ubiquitin-proteasome system.
  • PEG-based linker
  • Utilized in the synthesis of PROTACs
  • Connects ligands for E3 ubiquitin ligase and target proteins
  • Enables selective degradation of target proteins
  • Molecular weight: 413.30