Learn More
Medchemexpress LLC Bromoacetamido-PEG4-acid | 1807518-67-7 | MFCD28950779 | 96.8% | 386.24 g/mol | C13H24BrNO7 | 100 MG
SDP
Catalog No. 5000203768
Encompass_Preferred
Bromoacetamido-PEG4-acid is a PEG-based linker containing a bromoacetamido reactive group and a four-unit polyethylene glycol (PEG4) spacer. It is used in the synthesis of PROTAC molecules and as a cleavable linker for antibody-drug conjugates (ADCs). The compound is supplied as a colorless to light yellow liquid and is available in multiple package sizes with defined storage conditions.
Catalog No. 50-002-03768 Supplier Medchemexpress LLC Supplier No. HY141382100MG
Bromoacetamido-PEG4-acid is a PEG-based linker containing a bromoacetamido reactive group and a four-unit polyethylene glycol (PEG4) spacer. It is used in the synthesis of PROTAC molecules and as a cleavable linker for antibody-drug conjugates (ADCs). The compound is supplied as a colorless to light yellow liquid and is available in multiple package sizes with defined storage conditions.