Learn More
Medchemexpress LLC Bromoacetamido-PEG4-acid | 1807518-67-7 | MFCD28950779 | 96.8% | 386.24 g/mol | C13H24BrNO7 | 250 MG
SDP
Catalog No. 5000202681
Encompass_Preferred
Bromoacetamido-PEG4-acid is a PEG-based bifunctional linker containing a bromoacetamido reactive handle and a four-unit polyethylene glycol spacer terminating in a carboxylic acid. It is used in medicinal chemistry for constructing proteolysis targeting chimeras (PROTACs) and cleavable antibody-drug conjugate (ADC) linkers, providing a reactive site for conjugation and improved solubility.
  • Contains a bromoacetamido reactive handle for efficient conjugation.
  • Four-unit PEG spacer improves solubility and conformational flexibility.
  • Terminal carboxylic acid enables amide coupling and derivatization.
  • Supplied as a colorless to light yellow liquid with high purity.
  • Suitable for PROTAC and ADC synthesis in medicinal chemistry workflows.
Catalog No. 50-002-02681 Supplier Medchemexpress LLC Supplier No. HY141382250MG
Bromoacetamido-PEG4-acid is a PEG-based bifunctional linker containing a bromoacetamido reactive handle and a four-unit polyethylene glycol spacer terminating in a carboxylic acid. It is used in medicinal chemistry for constructing proteolysis targeting chimeras (PROTACs) and cleavable antibody-drug conjugate (ADC) linkers, providing a reactive site for conjugation and improved solubility.
  • Contains a bromoacetamido reactive handle for efficient conjugation.
  • Four-unit PEG spacer improves solubility and conformational flexibility.
  • Terminal carboxylic acid enables amide coupling and derivatization.
  • Supplied as a colorless to light yellow liquid with high purity.
  • Suitable for PROTAC and ADC synthesis in medicinal chemistry workflows.