Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bromoacetamido-PEG4-acid | 1807518-67-7 | MFCD28950779 | 96.8% | 386.24 g·mol⁻¹ | C13H24BrNO7 | 500 MG
SDP

Catalog No. 5000203766
Encompass_Preferred

Bromoacetamido-PEG4-acid is a 4-unit polyethylene glycol (PEG4) linker bearing a bromoacetamido functional group. It is used as a cleavable spacer and reactive handle in the synthesis of PROTACs and antibody-drug conjugates (ADCs), providing aqueous solubility and sites for nucleophilic conjugation.

  • 4-unit PEG spacer improves solubility and flexibility
  • Bromoacetamido reactive group enables nucleophilic substitution conjugation
  • Molecular weight 386.24 g·mol⁻¹
  • CAS number 1807518-67-7
  • Purity 96.8% (as supplied)
  • Typical package size 500 MG

Catalog No. 50-002-03766 Supplier Medchemexpress LLC Supplier No. HY141382500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bromoacetamido-PEG4-acid is a 4-unit polyethylene glycol (PEG4) linker bearing a bromoacetamido functional group. It is used as a cleavable spacer and reactive handle in the synthesis of PROTACs and antibody-drug conjugates (ADCs), providing aqueous solubility and sites for nucleophilic conjugation.

  • 4-unit PEG spacer improves solubility and flexibility
  • Bromoacetamido reactive group enables nucleophilic substitution conjugation
  • Molecular weight 386.24 g·mol⁻¹
  • CAS number 1807518-67-7
  • Purity 96.8% (as supplied)
  • Typical package size 500 MG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.