Learn More
Medchemexpress LLC Bromoacetamido-PEG4-C2-Boc | 1807521-00-1 | 1 G
SDP
Catalog No. 5000340331
Encompass_Preferred
Bromoacetamido-PEG4-C2-Boc is a laboratory chemical used for the manufacture of substances. It is stable under recommended storage conditions.
  • Harmful if swallowed
  • Causes skin and serious eye irritation
  • May cause respiratory irritation
  • Store in a cool, well-ventilated area away from direct sunlight and ignition sources
  • Recommended storage for pure form: -20°C for 3 years or 4°C for 2 years
  • Recommended storage for in solvent: -80°C for 6 months or -20°C for 1 month
Catalog No. 50-003-40331 Supplier Medchemexpress LLC Supplier No. HY1305201G
Bromoacetamido-PEG4-C2-Boc is a laboratory chemical used for the manufacture of substances. It is stable under recommended storage conditions.
  • Harmful if swallowed
  • Causes skin and serious eye irritation
  • May cause respiratory irritation
  • Store in a cool, well-ventilated area away from direct sunlight and ignition sources
  • Recommended storage for pure form: -20°C for 3 years or 4°C for 2 years
  • Recommended storage for in solvent: -80°C for 6 months or -20°C for 1 month