Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bromoacetamido-PEG4-C2-Boc | 1807521-00-1 | 250 MG
SDP

Catalog No. 5000341966
Encompass_Preferred

Bromoacetamido-PEG4-C2-Boc is a laboratory chemical (CAS No. 1807521-00-1) primarily utilized in the manufacture of substances. This compound exhibits good stability when stored under recommended conditions and typically appears as a liquid. It possesses a molecular formula of C17H32BrNO7 and a molecular weight of 442.34.

  • Suitable for laboratory research and substance manufacturing
  • Stable under recommended storage conditions
  • Appears as a liquid
  • Molecular formula: C17H32BrNO7
  • Molecular weight: 442.34
  • Storage (pure form): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month
  • Shipping at room temperature if less than 2 weeks

Catalog No. 50-003-41966 Supplier Medchemexpress LLC Supplier No. 250MGHY130520
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bromoacetamido-PEG4-C2-Boc is a laboratory chemical (CAS No. 1807521-00-1) primarily utilized in the manufacture of substances. This compound exhibits good stability when stored under recommended conditions and typically appears as a liquid. It possesses a molecular formula of C17H32BrNO7 and a molecular weight of 442.34.

  • Suitable for laboratory research and substance manufacturing
  • Stable under recommended storage conditions
  • Appears as a liquid
  • Molecular formula: C17H32BrNO7
  • Molecular weight: 442.34
  • Storage (pure form): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month
  • Shipping at room temperature if less than 2 weeks

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.