Learn More
Medchemexpress LLC Bromoacetamido-PEG4-NHS ester | 1260139-70-5 | 98.69% | C17H27BrN2O9 | 250 MG
SDP
Catalog No. 5000357341
Encompass_Preferred
Bromoacetamido-PEG4-NHS ester is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs consist of two distinct ligands joined by a linker; one ligand targets an E3 ubiquitin ligase, and the other targets the protein of interest. PROTACs function by leveraging the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PROTAC linker
  • Suitable for the synthesis of PROTACs
  • Intended for research use only
Catalog No. 50-003-57341 Supplier Medchemexpress LLC Supplier No. 250MGHY141387
Bromoacetamido-PEG4-NHS ester is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs consist of two distinct ligands joined by a linker; one ligand targets an E3 ubiquitin ligase, and the other targets the protein of interest. PROTACs function by leveraging the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PROTAC linker
  • Suitable for the synthesis of PROTACs
  • Intended for research use only