Learn More
Medchemexpress LLC Bromoacetamido-peg4-nhs ester | 1260139-70-5 | 98.7% | C17H27BrN2O9 | 100 MG
SDP
Catalog No. 5000356832
Encompass_Preferred
Bromoacetamido-PEG4-NHS ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. These PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-56832 Supplier Medchemexpress LLC Supplier No. HY141387100MG
Bromoacetamido-PEG4-NHS ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. These PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Selectively degrades target proteins