Learn More
Medchemexpress LLC Bromoacetamido-PEG4-NHS ester | 1260139-70-5 | 98.7% | C17H27BrN2O9 | 50 MG
SDP
Catalog No. 5000356913
Encompass_Preferred
Bromoacetamido-PEG4-NHS ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Enables selective degradation of target proteins
  • Utilizes the intracellular ubiquitin-proteasome system
Catalog No. 50-003-56913 Supplier Medchemexpress LLC Supplier No. HY14138750MG
Bromoacetamido-PEG4-NHS ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Enables selective degradation of target proteins
  • Utilizes the intracellular ubiquitin-proteasome system