Learn More
Medchemexpress LLC Bromoacetamido-PEG5-azide | 1415800-37-1 | 95.5% | C14H27BrN4O6 | 10MG
SDP
Catalog No. 5000261405
Encompass_Preferred
Bromoacetamido-PEG5-azide is a heterobifunctional PEG-based linker used for PROTAC assembly and bioconjugation. It combines a bromoacetamido electrophile, which undergoes nucleophilic substitution with thiols and other nucleophiles, with an azide group that participates in copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted azide-alkyne cycloaddition (SPAAC). The PEG5 spacer imparts flexibility and improved solubility, and the material is supplied at approximately 95.5% purity for research applications.
  • Heterobifunctional design enables sequential orthogonal reactions.
  • Bromoacetamido group reacts selectively with nucleophiles, such as thiols.
  • Azide moiety compatible with CuAAC and SPAAC click chemistries.
  • PEG5 spacer enhances solubility and conformational flexibility.
  • Reported high purity suitable for research and synthesis workflows.
  • Available in research-scale quantities for method development.
Catalog No. 50-002-61405 Supplier Medchemexpress LLC Supplier No. HY14082610MG
Bromoacetamido-PEG5-azide is a heterobifunctional PEG-based linker used for PROTAC assembly and bioconjugation. It combines a bromoacetamido electrophile, which undergoes nucleophilic substitution with thiols and other nucleophiles, with an azide group that participates in copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted azide-alkyne cycloaddition (SPAAC). The PEG5 spacer imparts flexibility and improved solubility, and the material is supplied at approximately 95.5% purity for research applications.
  • Heterobifunctional design enables sequential orthogonal reactions.
  • Bromoacetamido group reacts selectively with nucleophiles, such as thiols.
  • Azide moiety compatible with CuAAC and SPAAC click chemistries.
  • PEG5 spacer enhances solubility and conformational flexibility.
  • Reported high purity suitable for research and synthesis workflows.
  • Available in research-scale quantities for method development.