Learn More
Medchemexpress LLC Bromoacetamido-PEG5-DOTA | 2353410-19-0 | 98.0% | C30H55BrN6O13 | 50 MG
SDP
Catalog No. 5000353793
Encompass_Preferred
Bromoacetamido-PEG5-DOTA is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. These compounds employ two distinct ligands connected by a linker, one targeting an E3 ubiquitin ligase and the other a target protein. PROTACs leverage the intracellular ubiquitin-proteasome system for selective degradation of target proteins. This product appears as a white to off-white oil.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Appears as a white to off-white oil
  • For research use only
Catalog No. 50-003-53793 Supplier Medchemexpress LLC Supplier No. HY14075450MG
Bromoacetamido-PEG5-DOTA is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. These compounds employ two distinct ligands connected by a linker, one targeting an E3 ubiquitin ligase and the other a target protein. PROTACs leverage the intracellular ubiquitin-proteasome system for selective degradation of target proteins. This product appears as a white to off-white oil.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Appears as a white to off-white oil
  • For research use only