Learn More
Medchemexpress LLC Bromoacetamido-PEG5-DOTA | 2353410-19-0 | 98.02% | C30H55BrN6O13 | 25 MG
SDP
Catalog No. 5000352817
Encompass_Preferred
Bromoacetamido-PEG5-DOTA is a PEG-based PROTAC linker crucial for synthesizing PROTACs. These molecules employ the intracellular ubiquitin-proteasome system to selectively degrade target proteins by linking an E3 ubiquitin ligase ligand with a target protein ligand. It appears as a white to off-white oil and is exclusively for research purposes.
  • PEG-based PROTAC linker
  • Used in PROTAC synthesis
  • Appears as an oil
  • Color ranges from white to off-white
  • Exclusively for research use
Catalog No. 50-003-52817 Supplier Medchemexpress LLC Supplier No. 25MGHY140754
Bromoacetamido-PEG5-DOTA is a PEG-based PROTAC linker crucial for synthesizing PROTACs. These molecules employ the intracellular ubiquitin-proteasome system to selectively degrade target proteins by linking an E3 ubiquitin ligase ligand with a target protein ligand. It appears as a white to off-white oil and is exclusively for research purposes.
  • PEG-based PROTAC linker
  • Used in PROTAC synthesis
  • Appears as an oil
  • Color ranges from white to off-white
  • Exclusively for research use