Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromoacetic acid, 99%, MP Biomedicals™

Catalog No. ICN19382380 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
ICN19382380 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19382380 Supplier MP Biomedicals, Inc Supplier No. 0219382380

missing translation for 'validMainframeMsg'

Specifications

CAS 79-08-3
Color Orange to White
Assay Percent Range 0.99
Molecular Formula C2H3BrO2
Quantity 100 g
Synonym bromoacetic acid, monobromoacetic acid, bromoethanoic acid, acetic acid, bromo, to ntu, bromoacetate ion, acide bromacetique, 2-bromoacetyl group, 2-bromoethanoic acid, acetic acid, 2-bromo
InChI Key KDPAWGWELVVRCH-UHFFFAOYSA-N
SMILES C(C(=O)O)Br
IUPAC Name 2-bromoacetic acid
Molecular Weight (g/mol) 138.948
PubChem CID 6227
Formula Weight 138.9
Physical Form Solid
Chemical Name or Material Bromoacetic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.