Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bromoacetic Acid N-Hydroxysuccinimide Ester

Catalog No. B8271100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B8271100MG 100 mg
B82711G 1 g
B8271250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B8271100MG Supplier Sigma Aldrich Supplier No. B8271100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

A heterobifunctional cross-linking reagent which allows bromoacetylation of primary amine groups followed by coupling to sulfhydryl-containing compounds. Typically, initial reaction couples via ester to primary amine by amide bond formation in the pH range 6.5-8.5. The second reaction results in thioether bonding in pH range 7.0-8.0.

Specifications

CAS 107-88-0
Density 1.005 g/mL at 25°C
Molecular Formula C4H10O2
Linear Formula CH3CH(OH)CH2CH2OH
Quantity 100 mg
MDL Number MFCD00004554
Molecular Weight (g/mol) 90.12 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.