Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bromoacetyl bromide

Catalog No. B564125G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B564125G 5 g
B56412100G 100 g
B56412500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B564125G Supplier Sigma Aldrich Supplier No. B564125G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Bromoacetyl bromide can be used to convert:
  • Amines to azido acetamides
  • P-arsanilic acid to 4-(2-bromoacetylamino)benzenearsonic acid, a precursor to 4-(n-(s-penicillaminylacetyl)amino)phenylarsonous acid (penao), a metal-based drug
  • 3,5-dimethylphenol to 3,5-dimethylphenyl 2-bromoacetate
  • Cystamine dihydrochloride to n,nα-bis(bromoacetyl) cystamine, a bifunctional quaternizing agent.it can also be used as a reagent to synthesize imidazolium or piridinium based ionic liquids

Specifications

CAS 598-21-0
Density 2.317 g/mL (at 25°C (literature))
Boiling Point 147°C to 150°C (lit.)
Molecular Formula C2H2Br2O
Refractive Index n20/D 1.547 (literature)
Linear Formula BrCH2COBr
MDL Number MFCD00000115
Quantity 5 g
Molecular Weight (g/mol) 201.84
Percent Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.