Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromoacetyl Bromide 98.0+%, TCI America™
SDP

Catalog No. B0539100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0539100G 100 g
B053925G 25 g
B0539500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B0539100G Supplier TCI America Supplier No. B0539100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 598-21-0
Molecular Formula C2H2Br2O
Molecular Weight (g/mol) 201.845
MDL Number MFCD00000115
InChI Key LSTRKXWIZZZYAS-UHFFFAOYSA-N
Synonym bromoacetyl bromide, acetyl bromide, bromo, bromoacetylbromide, acetyl bromide, 2-bromo, bromo acetyl bromide, ccris 9090, bromoacetic acid bromide, bromacetylbromide, bromacetyl bromide, bromoactyl bromide
PubChem CID 61147
IUPAC Name 2-bromoacetyl bromide
SMILES C(C(=O)Br)Br

Specifications

Color Yellow
Boiling Point 150°C
Quantity 100 g
UN Number 2513
Formula Weight 201.85
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Bromoacetyl Bromide
EINECSNumber (9)-1233&(9)-1275
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.