Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bromoacetyl chloride

Catalog No. 16120100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-120-100ML 100 mL
16-120-25ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16-120-100ML Supplier Sigma Aldrich Supplier No. 16120100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • The competitive photodissociation of bromoacetyl chloride has been studied by ab initio methods.bromoacetyl chloride was used in the synthesis of α,α-disubstituted thioisom++nchnones
  • It was also used in the preparation of 1,3-dibromoacetone

Specifications

CAS 22118-09-8
Density 1.89 g/mL (at 20°C)
Boiling Point 127°C to 128°C (lit.)
Molecular Formula C2H2BrClO
Refractive Index n20/D 1.495
Linear Formula BrCH2COCl
MDL Number MFCD00000724
Quantity 100 mL
Molecular Weight (g/mol) 157.39
Percent Purity ≥95% (GC)
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.