Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bromoacetyl-PEG3-DBCO | 00-00-0 | 98.6% | 600.50 | C29H34BrN3O6 | 5 MG
SDP

Catalog No. 5000262629
Encompass_Preferred

Bromoacetyl-PEG3-DBCO is a heterobifunctional PEG-based linker for bioconjugation and PROTAC synthesis. It combines a DBCO bioorthogonal handle with a bromoacetyl electrophile and a short PEG3 spacer to enable efficient, selective conjugation and good solubility in small-scale synthetic workflows.

  • Contains a DBCO moiety for strain-promoted alkyne-azide cycloaddition (SPAAC).
  • Provides a bromoacetyl reactive handle for selective conjugation to thiols.
  • PEG3 spacer enhances solubility and flexibility in conjugates.
  • High purity (~98.6%) suitable for synthetic applications.
  • Oil appearance facilitates handling for small-scale synthesis.
  • Recommended storage at -20°C in pure form for long-term stability.
  • Available in milligram-scale quantities for research use.

Catalog No. 50-002-62629 Supplier Medchemexpress LLC Supplier No. HY1334805MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bromoacetyl-PEG3-DBCO is a heterobifunctional PEG-based linker for bioconjugation and PROTAC synthesis. It combines a DBCO bioorthogonal handle with a bromoacetyl electrophile and a short PEG3 spacer to enable efficient, selective conjugation and good solubility in small-scale synthetic workflows.

  • Contains a DBCO moiety for strain-promoted alkyne-azide cycloaddition (SPAAC).
  • Provides a bromoacetyl reactive handle for selective conjugation to thiols.
  • PEG3 spacer enhances solubility and flexibility in conjugates.
  • High purity (~98.6%) suitable for synthetic applications.
  • Oil appearance facilitates handling for small-scale synthesis.
  • Recommended storage at -20°C in pure form for long-term stability.
  • Available in milligram-scale quantities for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.