Learn More
Medchemexpress LLC Bromoacetyl-PEG3-DBCO | 00-00-0 | 98.6% | C29H34BrN3O6 | 10MG
SDP
Catalog No. 5000261420
Encompass_Preferred
Bromoacetyl-PEG3-DBCO is a heterobifunctional PEG linker containing a dibenzocyclooctyne (DBCO) moiety and a bromoacetyl reactive handle. It is designed for conjugation and linker installation in PROTAC and bioconjugation workflows, enabling click chemistry and selective nucleophile coupling.
  • Facilitates SPAAC via a strained alkyne DBCO group.
  • Provides a bromoacetyl handle for selective conjugation to thiols or amines.
  • PEG3 spacer improves solubility and reduces steric hindrance.
  • High purity suitable for research and synthesis workflows.
  • Available in small-scale pack sizes for discovery chemistry.
Catalog No. 50-002-61420 Supplier Medchemexpress LLC Supplier No. HY13348010MG
Bromoacetyl-PEG3-DBCO is a heterobifunctional PEG linker containing a dibenzocyclooctyne (DBCO) moiety and a bromoacetyl reactive handle. It is designed for conjugation and linker installation in PROTAC and bioconjugation workflows, enabling click chemistry and selective nucleophile coupling.
  • Facilitates SPAAC via a strained alkyne DBCO group.
  • Provides a bromoacetyl handle for selective conjugation to thiols or amines.
  • PEG3 spacer improves solubility and reduces steric hindrance.
  • High purity suitable for research and synthesis workflows.
  • Available in small-scale pack sizes for discovery chemistry.