Learn More
American Radiolabeled Chemicals Inc BROMOBENZ R-3H 1LC
SDP
Catalog No. 50352909
Encompass_Preferred
Bromobenzene-ring-2,4,6-3H 1mCi in ethanol, 1 mCi/ml, 70-80 Ci/mmol, M.W. 157.01, Shipped in Dry Ice, Exclusive
Catalog No. 50-352-909 Supplier American Radiolabeled Chemicals Inc Supplier No. ART 19661MC
Bromobenzene-ring-2,4,6-3H 1mCi in ethanol, 1 mCi/ml, 70-80 Ci/mmol, M.W. 157.01, Shipped in Dry Ice, Exclusive