Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromobenzene (Certified), Fisher Chemical

Catalog No. b2531 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
B253-1 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B253-1
This item is not returnable. View return policy

Chemical Identifiers

CAS 108-86-1
Molecular Formula C6H5Br
Molecular Weight (g/mol) 157.01
MDL Number MFCD00000055
InChI Key QARVLSVVCXYDNA-UHFFFAOYSA-N
Synonym benzene, bromo, monobromobenzene, phenyl bromide, bromo-benzene, 1-bromobenzene, phbr, bromo benzene, 4-bromobenzene, unii-co4d5j547l, hsdb 47
PubChem CID 7961
ChEBI CHEBI:3179
IUPAC Name bromobenzene
SMILES BrC1=CC=CC=C1

Specifications

Boiling Range 2°C max.
Melting Point -31°C
Density 1.490g/cm³
Boiling Point 156°C
Flash Point 51°C
Packaging Glass Bottle
Quantity 1 L
UN Number 2514
Formula Weight 157.01
Grade Certified
Physical Form Liquid
Chemical Name or Material Bromobenzene
Show More Show Less
DOTInformation Class 3: Flammable Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.