Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromobenzene-d{5}, 99% (Isotopic)

Catalog No. AA4226909
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4226909 10 g
AA4226904 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4226909 Supplier Thermo Scientific Chemicals Supplier No. 04226909
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 4165-57-5 | C6H5Br | 162.04 g/mol

The labeled version of bromobenzene , used as a general reagent in palladium-catalyzed reaction and in the synthesize of Grignard reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4165-57-5
Molecular Formula C6H5Br
Molecular Weight (g/mol) 162.04
MDL Number MFCD00000056
InChI Key QARVLSVVCXYDNA-RALIUCGRSA-N
Synonym bromobenzene-d5, bromo 2h5 benzene, 1-bromo 2 h? benzene, bromobenzene d5, benzene-d5, bromo, 1-bromo 2h5 benzene, c6d5br, bromobenzene-d5, 99.5 atom % d, bromobenzene-d5, reagent grade, ≥99 atom % d cp
PubChem CID 2723968
SMILES [2H]C1=C([2H])C([2H])=C(Br)C([2H])=C1[2H]

Specifications

Physical Form Liquid
Quantity 10 g
Density 1.55 g/mL
Flash Point 51°C (124°F)
Odor Aromatic
Refractive Index 1.557
Boiling Point 53°C (23mmHg)
UN Number UN2514
Solubility Information Insoluble in water.
IUPAC Name 1-bromo-2,3,4,5,6-pentadeuteriobenzene
Formula Weight 162.06
Percent Purity 99%
Chemical Name or Material Bromobenzene-d5, Isotopic
Show More Show Less
Hazard Category H226-H315-H319
Hazard Statement GHS H Statement
H226-H315
Flammable liquid and vapor.
Causes skin irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: BROMOBENZENE
EINECSNumber 224-013-8
TSCA No
Recommended Storage Ambient

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.