Learn More
Medchemexpress LLC Bromobenzene-d5 | 4165-57-5 | MFCD00000056 | 25g
SDP
Catalog No. 5000266355
Encompass_Preferred
Bromobenzene-d5 is the deuterium labeled Bromobenzene Bromobenzene is a well-known environmental toxin which causes liver and kidney damage through CYP450-mediated bio-activation to generate reactive metabolites and consequently oxidative stress[1]
Catalog No. 50-002-66355 Supplier Medchemexpress LLC Supplier No. HYZ0056S25G
Bromobenzene-d5 is the deuterium labeled Bromobenzene Bromobenzene is a well-known environmental toxin which causes liver and kidney damage through CYP450-mediated bio-activation to generate reactive metabolites and consequently oxidative stress[1]