Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromobenzene-d5, for NMR, 99.5 atom% D

Catalog No. AC321230100
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC321230100 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC321230100 Supplier Thermo Scientific Chemicals Supplier No. 321230100
Add to Cart

CAS: 4165-57-5 | C6H5Br | 162.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4165-57-5
Molecular Formula C6H5Br
Molecular Weight (g/mol) 162.04
MDL Number MFCD00000056
InChI Key QARVLSVVCXYDNA-RALIUCGRSA-N
IUPAC Name 1-bromo(²H₅)benzene
SMILES [2H]C1=C([2H])C([2H])=C(Br)C([2H])=C1[2H]

Specifications

Density 1.5500 g/mL
Quantity 10 mL
Refractive Index 1.5568
Specific Gravity 1.55
Flash Point 51°C
Boiling Point 53.0°C (23.0 mmHg)
Linear Formula C6D5Br
Beilstein 05, III, 561
Solubility Information Solubility aq. soln.: insoluble.
Formula Weight 162.06
Grade NMR
Chemical Name or Material Bromobenzene-d5
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation.

Toxic to aquatic life with long lasting effects.

Flammable liquid and vapor.

Precautionary Statement GHS P Statement Avoid release to the environment.

IF ON SKIN: Wash with plenty of soap and water.

Keep away from heat, hot surfaces, sparks, open flames and other ignition sources.

No smoking.

EINECSNumber 224-013-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.