Learn More
Medchemexpress LLC HY-100041 25mg Medchemexpress, Bromobimane CAS:71418-44-5 Purity:>98%
SDP
Catalog No. 501931708
Encompass_Preferred
Medchemexpress, HY-100041 25mg Bromobimane CAS:71418-44-5 Bromobimane is essentially nonfluorescent and converts into fluorescent products when reacts with small thiols. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1708 Supplier Medchemexpress LLC Supplier No. HY10004125MG
Medchemexpress, HY-100041 25mg Bromobimane CAS:71418-44-5 Bromobimane is essentially nonfluorescent and converts into fluorescent products when reacts with small thiols. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.