Learn More
Medchemexpress LLC Bromobimane | 71418-44-5 | MFCD00036951 | 99.3% | 271.11 g/mol | C10H11BrN2O2 | 100 MG
SDP
Catalog No. 5000215781
Encompass_Preferred
Bromobimane is a thiol-reactive fluorescent labeling reagent that is nonfluorescent until it reacts with thiol groups to form fluorescent adducts. It is commonly used to derivatize and detect thiol-containing molecules in biochemical and analytical workflows, including HPLC and fluorescence-based assays.
  • Selective reactivity with thiol groups.
  • Nonfluorescent until formation of fluorescent adducts.
  • Excitation 344 nm, emission 450 nm for fluorescence detection.
  • High purity (99.3%) suitable for analytical use.
  • Compatible with HPLC and fluorescence quantification methods.
  • Available as solid and as DMSO solution for convenience.
Catalog No. 50-002-15781 Supplier Medchemexpress LLC Supplier No. HY100041100MG
Bromobimane is a thiol-reactive fluorescent labeling reagent that is nonfluorescent until it reacts with thiol groups to form fluorescent adducts. It is commonly used to derivatize and detect thiol-containing molecules in biochemical and analytical workflows, including HPLC and fluorescence-based assays.
  • Selective reactivity with thiol groups.
  • Nonfluorescent until formation of fluorescent adducts.
  • Excitation 344 nm, emission 450 nm for fluorescence detection.
  • High purity (99.3%) suitable for analytical use.
  • Compatible with HPLC and fluorescence quantification methods.
  • Available as solid and as DMSO solution for convenience.