Learn More
Medchemexpress LLC Bromobimane | 71418-44-5 | MFCD00036951 | 99.3% | 271.11 g/mol | C10H11BrN2O2 | 50 MG
SDP
Catalog No. 5000214984
Encompass_Preferred
Bromobimane is a thiol-reactive fluorescent dye that is essentially nonfluorescent until it alkylates thiol groups to form strongly fluorescent adducts. It is used to label, detect, and quantify thiols in biochemical and analytical workflows.
  • Reacts selectively with thiol groups to generate fluorescent products.
  • Fluorescent properties: excitation 344 nm, emission 450 nm.
  • High chemical purity suitable for sensitive assays.
  • Compatible with common fluorescence detection methods.
  • Supplied for laboratory research applications.
Catalog No. 50-002-14984 Supplier Medchemexpress LLC Supplier No. HY10004150MG
Bromobimane is a thiol-reactive fluorescent dye that is essentially nonfluorescent until it alkylates thiol groups to form strongly fluorescent adducts. It is used to label, detect, and quantify thiols in biochemical and analytical workflows.
  • Reacts selectively with thiol groups to generate fluorescent products.
  • Fluorescent properties: excitation 344 nm, emission 450 nm.
  • High chemical purity suitable for sensitive assays.
  • Compatible with common fluorescence detection methods.
  • Supplied for laboratory research applications.