Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromobimane 98.0+%, TCI America™
SDP

Catalog No. B422020MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
20 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B422020MG 20 mg
B4220100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B422020MG Supplier TCI America Supplier No. B422020MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 71418-44-5
Molecular Formula C10H11BrN2O2
Molecular Weight (g/mol) 271.11
MDL Number MFCD00036951
InChI Key AHEWZZJEDQVLOP-UHFFFAOYSA-N
Synonym Monobromobimane, 3-(Bromomethyl)-2,5,6-trimethylpyrazolo[1,2-a]pyrazole-1,7-dione
PubChem CID 114810
ChEBI CHEBI:62825
IUPAC Name 3-(bromomethyl)-2,5,6-trimethyl-1H,7H-[1,2]diazolo[1,2-a]pyrazole-1,7-dione
SMILES CC1=C(C)C(=O)N2N1C(CBr)=C(C)C2=O

Specifications

Melting Point 162°C
Color Yellow
Quantity 20 mg
Formula Weight 271.11
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Bromobimane
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.