Learn More
Bioworld BromoBlue Universal Loading Dye (6X), 10 mL
Catalog No. 50196774
Encompass_Preferred
Premixed and ready-to-load gel dyes. Glow Loading Dyes generate intense DNA/RNA bands with little background or band distortion and can be used with any type of agarose or acrylamide gels. BromoBlue dye contains bromophenol blue in a Ficoll-based solution (6X concentration).
Catalog No. 50-196-774 Supplier Bioworld Supplier No. 105700051
Premixed and ready-to-load gel dyes. Glow Loading Dyes generate intense DNA/RNA bands with little background or band distortion and can be used with any type of agarose or acrylamide gels. BromoBlue dye contains bromophenol blue in a Ficoll-based solution (6X concentration).