Learn More
AdipoGen Bromobutide
SDP
Supplier:  AdipoGen CDXB0124M250
Encompass_Preferred
Chemical. CAS 74712-19-9. Formula C15H22BrNO. MW 312.3. Synthetic. Amide herbicide pigment inhibitor. Mode of action is through inhibition of 4-hydroxyphenyl-pyruvatedioxygenase 4-HPPD that causes bleaching. Compound can be used as analytical reference material.
Catalog No. 50-256-1668