Learn More
Qorpak™ Bromobutyl Stoppers for Serum Vials
Catalog No. 02930158
Change view
Click to view available options
Quantity:
1000/Cs.
200/Cs.
5000/Cs.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-930-158 200/Cs.
02-930-159 1000/Cs.
02-930-154 5000/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02-930-158 Supplier Qorpak™ Supplier No. 345417
Only null left
Add to Cart
Add to Cart
Bromobutyl stoppers are high-performance elastomeric closures, crucial in pharma and labs. Ideal for sensitive products like lyophilized drugs and anaerobic cultures in vials or bottles.
  • Excellent gas impermeability
  • Superior self-sealing after syringe entry
  • Excellent chemical resistance
  • Suitability for autoclaving
  • Low particulates
Specifications
Closure Color Gray
Diameter (Metric) Outer 20 mm
Material Bromobutyl
Pre-slit No
Sterility Non-sterile
Disposable Limited-use
Graduated No
Closure Material Bromobutyl
Quantity 200/Cs.
Closure Size 20 mm
Marking Spot No
Autoclavable Autoclavable