Learn More
AdipoGen Bromocamphor-sulfonic acid H2O
SDP
Catalog No. 502561657
Encompass_Preferred
Chemical. CAS 206860-46-0. Formula C10H15BrO4S . xH2O. MW 311.19 anhydrous basis. Synthetic Chiral reagent used for the resolution of bases. Building block for synthesis.
Catalog No. 50-256-1657 Supplier AdipoGen Supplier No. CDXB0135G001
Chemical. CAS 206860-46-0. Formula C10H15BrO4S . xH2O. MW 311.19 anhydrous basis. Synthetic Chiral reagent used for the resolution of bases. Building block for synthesis.