Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromochloromethane, MilliporeSigma™ Supelco™

Catalog No. 111018822 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8822 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8822 Supplier MilliporeSigma Supelco Supplier No. 442498
Add to Cart
Add to Cart

Sigma-Aldrich offers bromochloromethane for a broad-range applications such as environmental, food and beverage, forensics, petrochemical and pharmaceutical analyzes.

Environmental
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
CAS 74-97-5
Vapor Pressure 117 mmHg (20°C)
Vapor Density 4.5 (vs. air)
Density 1.991 g/mL (at 25°C)
Physical Form Neat
Linear Formula BrCH2 Cl
MDL Number MFCD00000880
Melting Point -88°C
Boiling Point 68°C
Type Organic Pollutant Standard
Quantity 500 mg
Formula Weight 129.38
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.