Learn More
Medchemexpress LLC Bromocholine bromide | 2758-06-7 | 98.0% | 246.97 | 5 G
SDP
Catalog No. 5000453036
Encompass_Preferred
2-Bromo-N,N,N-trimethylethanaminium bromide is a quaternary ammonium salt consisting of a positively charged 2-bromo-N,N,N-trimethylethylamine cation and a negatively charged bromide anion. This compound is commonly used as a phase transfer catalyst in organic chemical reactions, facilitating the transfer of reactants between immiscible phases. It can also be used as a halogenating agent, for example in the bromination of olefins. For research use only. We do not sell to patients. Bromocholine bromide is also a biochemical reagent that can be used as a biological material or organic compound for life science related research.
  • Commonly used as a phase transfer catalyst in organic chemical reactions.
  • Facilitates the transfer of reactants between immiscible phases.
  • Can be used as a halogenating agent, for example in the bromination of olefins.
  • Biochemical reagent for life science related research.
Catalog No. 50-004-53036 Supplier Medchemexpress LLC Supplier No. HYW0119725G
2-Bromo-N,N,N-trimethylethanaminium bromide is a quaternary ammonium salt consisting of a positively charged 2-bromo-N,N,N-trimethylethylamine cation and a negatively charged bromide anion. This compound is commonly used as a phase transfer catalyst in organic chemical reactions, facilitating the transfer of reactants between immiscible phases. It can also be used as a halogenating agent, for example in the bromination of olefins. For research use only. We do not sell to patients. Bromocholine bromide is also a biochemical reagent that can be used as a biological material or organic compound for life science related research.
  • Commonly used as a phase transfer catalyst in organic chemical reactions.
  • Facilitates the transfer of reactants between immiscible phases.
  • Can be used as a halogenating agent, for example in the bromination of olefins.
  • Biochemical reagent for life science related research.